Merck
  • Effects of quartz addition on the mechanochemical dechlorination of chlorobiphenyl by using CaO.
Effects of quartz addition on the mechanochemical dechlorination of chlorobiphenyl by using CaO.
Environmental science & technology (2002-01-05)
Q Zhang, F Saito, T Ikoma, S Tero-Kubota, K Hatakeda
ABSTRACT
Grinding a mixture of 3-chlorobiphenyl (BP-Cl) and CaO with or without the addition of quartz was conducted in air by a planetary ball mill to investigate the mechanochemical dechlorination of BP-Cl. The dechlorinating reaction proceeds with an increase in grinding time, and over 99% of BP-Cl is decomposed at 360 min. Washing the ground sample with different solvents results in different products. Addition of quartz to the grinding mixture facilitates dechlorination efficiency, especially in the case of a high weight ratio of BP-Cl to CaO.
Product Description
Supelco
PCB No 2, analytical standard
Pricing and availability is not currently available.