Merck
  • Toxicity of synthetic piperonyl compounds to leaf-cutting ants and their symbiotic fungus.
Toxicity of synthetic piperonyl compounds to leaf-cutting ants and their symbiotic fungus.
Pest management science (2001-07-24)
S R Victor, F R Crisóstomo, F C Bueno, F C Pagnocca, J B Fernandes, A G Correa, O C Bueno, M J Hebling, M Bacci Júnior, P C Vieira, M F da Silva
ABSTRACT
The development of Leucoagaricus gongylophorus, the fungus cultured by the leaf-cutting ant Atta sexdens was inhibited in vitro by synthetic compounds containing the piperonyl group. In addition, worker ants that were fed daily on an artificial diet to which these compounds were added had a higher mortality rate than the controls. The inhibition of the fungal growth increased with the size of the carbon side chain ranging from C1 through C8 and decreasing thereafter. 1-(3,4-Methylenedioxybenzyloxy)octane (compound 5) was the most active compound and inhibited the fungal development by 80% at a concentration of 15 micrograms ml-1. With worker ants the toxic effects started with compound 5 and increased with the number of carbons in the side chain. Thus, for the same concentration (100 micrograms ml-1) the mortality rates observed after 8 days of diet ingestion were 82%, 66% and 42%, for 1-(3,4-methylenedioxybenzyloxy)decane, 1-(3,4-methylenedioxybenzyloxy)dodecane and compound 5, respectively, whereas with commercial piperonyl butoxide the mortality was 68%. The latter compound, which is known as a synergist insecticide, was as inhibitory to the symbiotic fungus as the synthetic compound 5. The possibility of controlling these insects in the future using compounds that can target simultaneously both organisms is discussed.
Sigma-Aldrich
Piperonyl alcohol, 98%