Merck
Intranasal diamorphine in children with trauma.
Emergency nurse : the journal of the RCN Accident and Emergency Nursing Association (2013-03-23)
Hannah Skuse, Vanessa Lawlor
ABSTRACT
This article discusses the administration of intranasal diamorphine as an analgesia-in paediatric trauma care. The authors outline evidence for its use and discuss a recent multicentre safety study of an intranasal diamorphine product. In addition, they advise emergency department nurses on administering the analgesia, and offer guidelines for determining patient suitability and post-administration assessment.
Product Description
Supelco
Heroin solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Heroin, analytical standard, ≥98% (HPLC)