Merck
Slow chemical transmission between dorsal root ganglion neuron somata.
The European journal of neuroscience (2012-08-01)
Gabriela M Rozanski, Hyunhee Kim, Qi Li, Fiona K Wong, Elise F Stanley
ABSTRACT
Somatic sensory neuron somata are located within the dorsal root ganglia (DRG) and are mostly ensheathed by individual satellite glial cell sheets. It has been noted, however, that a subpopulation of these DRG somata are intimately associated, separated only by a single thin satellite glial cell membrane septum. We set out to test whether such neuron-glial cell-neuron trimers (NGlNs) are also linked functionally. The presence of NGlNs in chick DRGs was confirmed by electron microscopy. Selective satellite glial cell immunostains were identified and were used to image the inter-neuron septa in DRG frozen sections. We used a gentle, dispase-based enzymatic method to isolate chick and rat NGlNs in vitro for double patch clamp recordings. In the majority of pairs tested, an action potential-like stimulus train delivered to one soma resulted in a delayed, noisy and long-duration response in its idle partner. The response to a second stimulus train given minutes later was markedly facilitated. Both bidirectional and unidirectional transmission was observed between the paired neurons. Transmission was chemical and block by the general purinergic blocker suramin implicated ATP as a neurotransmitter. We conclude that the two neuronal somata in the NGlN can communicate by chemical transmission, which may involve a transglial, bi-synaptic pathway. This novel soma-to-soma transmission reflects a novel form of processing that may play a role in sensory disorders in the DRG and interneuron communication in the central nervous system.
Sigma-Aldrich
Suramin sodium salt, ≥98% (TLC)