Merck
  • Cellular folates prevent polyglutamation of 5, 10-dideazatetrahydrofolate. A novel mechanism of resistance to folate antimetabolites.
Cellular folates prevent polyglutamation of 5, 10-dideazatetrahydrofolate. A novel mechanism of resistance to folate antimetabolites.
The Journal of biological chemistry (1998-09-25)
A Tse, R G Moran
ABSTRACT
Mouse L1210 cell variants were selected for resistance to 5, 10-dideazatetrahydrofolate, a potent inhibitor of the first folate-dependent enzyme in de novo purine synthesis, glycinamide ribonucleotide formyltransferase. The drug-resistant phenotype selected was conditional to the folate compound used to support growth: grown on folic acid cells were 400-fold resistant, whereas they were 2.5-fold more sensitive to 5,10-dideazatetrahydrofolate than wild-type L1210 cells when grown on folinic acid. In folic acid-containing media, polyglutamation of 5, 10-dideazatetrahydrofolate was markedly reduced, yet folylpolyglutamate synthetase activity was not different from that in parental L1210 cells. Resistance was due to two changes in membrane transport: a minor increase in the Km for 5, 10-dideazatetrahydrofolate influx, and a major increase in folic acid transport. Enhanced folic acid transport resulted in an expanded cellular content of folates which blocked polyglutamation of 5,10-dideazatetrahydrofolate. We propose that polyglutamation of 5,10-dideazatetrahydrofolate is limited by feedback inhibition by cellular folates on folylpolyglutamate synthetase, an effect which reflects a mechanism in place to control the level of cellular folates. Although the primary alteration causative of resistance is different from those reported previously, all 5, 10-dideazatetrahydrofolate resistance phenotypes result in decreased drug polyglutamation, reflecting the centrality of this reaction to the action of 5,10-dideazatetrahydrofolate.
Sigma-Aldrich
Lometrexol hydrate, ≥95% (HPLC)