Merck
  • Automatic evaluation of antifungal volatile compounds on the basis of the dynamic growth process of a single hypha.
Automatic evaluation of antifungal volatile compounds on the basis of the dynamic growth process of a single hypha.
Applied microbiology and biotechnology (1993-03-01)
K Oh, H Matsuoka, O Sumita, K Takatori, H Kurata
ABSTRACT
An automatic analysing system was developed and employed for the evaluation of antifungal activity of volatile compounds in the gas phase. Aspergillus niger was inoculated on agar medium in the reaction vessel. The reaction vessel was incubated at 28 degrees C for 24 h and then a volatile compound was introduced into the vessel either in a batch or flow manner. The antifungal activity of the respective compounds estimated in situ was expressed by the dynamic response parameters of a single hypha. All volatiles tested in the present system inhibited hyphal growth, except linalyl acetate: Limone and geraniol were the most inhibitory. In contrast, linalyl acetate promoted hyphal growth. By definition of the parameters, the fungicidal and fungistatic effects could be distinguished.
Product Description
Supelco
Benzyl acetate, analytical standard
Sigma-Aldrich
Benzyl acetate, ≥99%
Sigma-Aldrich
Benzyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG