Merck
  • Prevention of scar spread on trunk excisions: a rater-blinded randomized controlled trial.
Prevention of scar spread on trunk excisions: a rater-blinded randomized controlled trial.
JAMA dermatology (2013-06-12)
Kevin F Kia, Molly V Burns, Travis Vandergriff, Sarah Weitzul
ABSTRACT
Wounds that heal under tension lead to wider and more conspicuous scars and result in decreased long-term patient satisfaction. We hypothesized that prolonged intradermal suture lifetime can decrease scar spread in wounds under tension. To determine whether prolonged intradermal support would help decrease scar spread. DESIGN Prospective, randomized, controlled, rater-blinded, split-scar trial. Outpatient dermatology clinic at Dallas Veterans Affairs Hospital, Dallas, Texas. Patients presenting with skin cancer on the trunk were considered for the trial. We included 25 distinct surgical sites on a total of 22 patients. After excision, the wounds were closed with polyglactin 910 and poly-4 hydroxybutyrate (P4HB) sutures in opposite halves of the same wound. Quantitative scar spread at 12 months and qualitative assessment using a visual analog scale and Hollander Wound Evaluation Scale. We found a statistically significant difference in scar width between the 2 suture materials, with a mean difference of 2.3 (95% CI, 1.0-3.6) mm (P < .001) favoring P4HB. A clinically significant difference on the visual analog and Hollander Wound Evaluation scales was not identified. Suture reactions were more common with P4HB. Prolonged intradermal suture support leads to significantly decreased scar spread. However, the use of a longer-acting absorbable suture increases the rate of suture reaction noted at 3 months. Further studies into less reactive, longer-acting biomaterials are needed. In clinical practice, excisions in high-tension areas that are classically known to spread over time can benefit from longer-acting intradermal sutures. TRIAL REGISTRATION clinicaltrials.gov Identifier: NCT00938691.
Sigma-Aldrich
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 502 H, Poly(D,L-lactide-co-glycolide), acid terminated, viscosity 0.16-0.24 dL/g 
Sigma-Aldrich
Resomer® RG 502, Poly(D,L-Lactide-co-Glycolide), lactide:glycolide 50:50, ester terminated, Mw 7,000-17,000
Sigma-Aldrich
Resomer® RG 752 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 75:25, Mw 4,000-15,000
Sigma-Aldrich
Resomer® RG 653 H, Poly(D,L-lactide-co-glycolide), acid terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 756 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 75:25, Mw 76,000-115,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), ester terminated, Mw 50,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide 65:35, Mw 40,000-75,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (75:25), mol wt 66,000-107,000
Sigma-Aldrich
Poly(D,L-lactide-co-glycolide), lactide:glycolide (50:50), mol wt 30,000-60,000
Sigma-Aldrich
Resomer® RG 504, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 858 S, Poly(D,L-lactide-co-glycolide), ester terminated, lactide:glycolide 85:15, Mw 190,000-240,000
Sigma-Aldrich
Resomer® RG 503, Poly(D,L-lactide-co-glycolide), lactide:glycolide 50:50, ester terminated, Mw 24,000-38,000
Sigma-Aldrich
Resomer® RG 504 H, Poly(D,L-lactide-co-glycolide), acid terminated, lactide:glycolide 50:50, Mw 38,000-54,000
Sigma-Aldrich
Resomer® RG 505, Poly(D,L-lactide-co-glycolide), ester terminated, Mw 54,000-69,000