Merck
  • Screening for long-lived genes identifies Oga1, a guanine-quadruplex associated protein that affects the chronological lifespan of the fission yeast Schizosaccharomyces pombe.
Screening for long-lived genes identifies Oga1, a guanine-quadruplex associated protein that affects the chronological lifespan of the fission yeast Schizosaccharomyces pombe.
Molecular genetics and genomics : MGG (2013-05-04)
Hokuto Ohtsuka, Shingo Ogawa, Hideaki Kawamura, Erika Sakai, Keiko Ichinose, Hiroshi Murakami, Hirofumi Aiba
ABSTRACT
Schizosaccharomyces pombe and Saccharomyces cerevisiae are excellent model organisms to study lifespan. We conducted screening to identify novel genes that, when overexpressed, extended the chronological lifespan of fission yeast. We identified seven genes, among which we focused on SPBC16A3.08c. The gene product showed similarity to Ylr150w of S. cerevisiae, which has affinity for guanine-quadruplex nucleic acids (G4). The SPBC16A3.08c product associated with G4 in vitro and complemented the phenotype of an S. cerevisiae Ylr150w deletion mutant. From these results, we proposed that SPBC16A3.08c encoded for a functional homolog of Ylr150w, which we designated ortholog of G4-associated protein (oga1 (+)). oga1 (+) overexpression extended the chronological lifespan and also decreased mating efficiency and caused both high and low temperature-sensitive growth. Deleting oga1 (+) resulted in caffeine-sensitive and canavanine-resistant phenotypes. Based on these results, we discuss the function of Oga1 on the chronological lifespan of fission yeast.
Product Description
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Sigma-Aldrich
L-Canavanine, ≥98% (TLC), powder, from Canavalia ensiformis
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, SAJ special grade, ≥98.5%
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine, European Pharmacopoeia (EP) Reference Standard