Merck
  • Efficient enantioselective synthesis of (R)-[3,5-bis(trifluoromethyl)phenyl] ethanol by Leifsonia xyli CCTCC M 2010241 using isopropanol as co-substrate.
Efficient enantioselective synthesis of (R)-[3,5-bis(trifluoromethyl)phenyl] ethanol by Leifsonia xyli CCTCC M 2010241 using isopropanol as co-substrate.
Journal of microbiology and biotechnology (2013-03-07)
Qi Ouyang, Pu Wang, Jin Huang, Jinbo Cai, Junyao He
ABSTRACT
(R)-[3,5-Bis(trifluoromethyl)phenyl] ethanol is a key chiral intermediate for the synthesis of aprepitant. In this paper, an efficient synthetic process for (R)-[3,5- bis(trifluoromethyl)phenyl] ethanol was developed via the asymmetric reduction of 3,5-bis(trifluoromethyl) acetophenone, catalyzed by Leifsonia xyli CCTCC M 2010241 cells using isopropanol as the co-substrate for cofactor recycling. Firstly, the substrate and product solubility and cell membrane permeability of biocatalysts were evaluated with different co-substrate additions into the reaction system, in which isopropanol manifested as the best hydrogen donor of coupled NADH regeneration during the bioreduction of 3,5-bis(trifluoromethyl) acetophenone. Subsequently, the optimization of parameters for the bioreduction were undertaken to improve the effectiveness of the process. The determined efficient reaction system contained 200 mM of 3,5-bis(trifluoromethyl) acetophenone, 20% (v/v) of isopropanol, and 300 g/l of wet cells. The bioreduction was executed at 30°C and 200 rpm for 30 h, and 91.8% of product yield with 99.9% of enantiometric excess (e.e.) was obtained. The established bioreduction reaction system could tolerate higher substrate concentrations of 3,5- bis(trifluoromethyl) acetophenone, and afforded a satisfactory yield and excellent product e.e. for the desired (R)-chiral alcohol, thus providing an alternative to the chemical synthesis of (R)-[3,5-bis(trifluoromethyl)phenyl] ethanol.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Pricing and availability is not currently available.
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Supelco
2-Propanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Isopropanol, 70% in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Pricing and availability is not currently available.