Merck
  • Efficacy and safety of mercuric oxide in the treatment of bacterial blepharitis.
Efficacy and safety of mercuric oxide in the treatment of bacterial blepharitis.
Antimicrobial agents and chemotherapy (1990-04-01)
R A Hyndiuk, E M Burd, A Hartz
ABSTRACT
A double-masked, placebo-controlled, randomized study was done to assess the safety and clinical and quantitative microbiologic efficacy of 1% mercuric oxide (yellow) ophthalmic ointment in the treatment of eyelid infections, i.e., bacterial blepharitis. A total of 39 patients with bacterial counts and clinical signs indicative of eyelid infection were treated twice daily for 7 days. Clinical biomicroscopic examination and quantitative microbiologic cultures were done just prior to initiation of treatment and again on days 3 and 7. Statistical analysis revealed a significant improvement in the clinical signs, bacterial count, cure rate, and improvement rate for subjects taking the active medication, compared with those taking the placebo on days 3 and 7. In addition, the medication was well tolerated.
Sigma-Aldrich
Mercury(II) oxide, 99.999% trace metals basis
Sigma-Aldrich
Mercury(II) oxide red, ACS reagent, ≥99.0%