Merck
  • Bacterial colonization and resistance patterns in 133 patients undergoing a primary hip- or knee replacement in Southern Sweden.
Bacterial colonization and resistance patterns in 133 patients undergoing a primary hip- or knee replacement in Southern Sweden.
Acta orthopaedica (2013-02-16)
Anna Stefánsdóttir, Asa Johansson, Lars Lidgren, Philippe Wagner, Annette W-Dahl
ABSTRACT
Prosthetic joint infections can be caused by bacteria derived from the patient's skin. The aim of the study was: (1) to determine which bacteria colonize the nose and groin in patients planned for primary hip or knee arthroplasty, (2) to determine the antimicrobial resistance patterns, and (3) to monitor changes in bacterial colonization and resistance patterns connected to surgery. 2 weeks before scheduled primary hip or knee arthroplasty, culture samples were taken from the anterior nares and from the groin of 133 consecutive patients. At surgery, cloxacillin was given prophylactically and cement with gentamicin was used. 2 weeks after surgery, another set of samples were taken from 120 of these patients. Bacterial findings and resistance patterns were analyzed. Preoperatively, 95% of the patients had coagulase-negative staphylococci (CNS) in the groin and 77% in the nose. The proportion of patients with a methicillin-resistant CNS in the groin increased from 20% preoperatively to 50% postoperatively (p < 0.001), and the proportion of patients with a gentamicin-resistant CNS in the groin increased from 5% to 45% (p < 0.001). 28% of the patients had Staphylococcus aureus in the nose preoperatively, and 7% in the groin. Methicillin-resistant Staphylococcus aureus (MRSA) was found in the nose of 1 patient. In southern Sweden, beta-lactams were effective against 99% of the Staphylococcus aureus strains and 80% of the CNS strains colonizing the patients undergoing primary hip or knee arthroplasty. Gentamicin protects against most CNS strains in cemented primary joint replacements.
Sigma-Aldrich
Cloxacillin sodium salt monohydrate, β-lactamase-resistant antibiotic