Merck
  • Nano zero-valent iron impregnated on titanium dioxide nanotube array film for both oxidation and reduction of methyl orange.
Nano zero-valent iron impregnated on titanium dioxide nanotube array film for both oxidation and reduction of methyl orange.
Water research (2013-02-05)
Dong-Min Yun, Hyun-Hee Cho, Jun-Won Jang, Jae-Woo Park
ABSTRACT
Here, we demonstrated that nano zero-valent iron (nZVI) impregnated onto self-organized TiO(2) nanotube thin films exhibits both oxidation and reduction capacities in addition to the possible electron transfer from TiO(2) to nZVI. The TiO(2) nanotubes were synthesized by anodization of titanium foil in a two-electrode system. Amorphous TiO(2) (amTiO(2)) nanotubes were annealed at 450 °C for 1 h to produce crystalline TiO(2) (crTiO(2)) nanotubes. The nZVI particles were immobilized on the TiO(2) array film by direct borohydride reduction. Field emission scanning electron microscopy (FE-SEM) analysis of the crystalline TiO(2) nanotube with nZVI (nZVI/crTiO(2)) indicated that the nZVI particles with a mean particle diameter of 28.38 ± 11.81 nm were uniformly distributed onto entire crTiO(2) nanotube surface with a mean pore diameter of 75.24 ± 17.66 nm and a mean length of 40.07 μm. Environmental applicability of our proposed nZVI/TiO(2) nanotube thin films was tested for methyl orange (MO) degradation in the aqueous system with and without oxygen. Since oxygen could facilitate the nZVI oxidation and inhibit electron transfer from crTiO(2) to nZVI surface, MO degradation by nZVI/crTiO(2) in the presence of oxygen was significantly suppressed whereas nZVI/crTiO(2) in the absence of oxygen enhanced MO degradation. MO degradation rate by each sample without oxygen were in following order: nZVI/crTiO(2) (k(obs) = 0.311 min(-1)) > nZVI/amTiO(2) (k(obs) = 0.164 min(-1)) > crTiO(2) (k(obs) = 0.068 min(-1)). This result can be explained with a synergistic effect of the significant reduction by highly-dispersed nZVI particles on TiO(2) nanotubes as well as the electron transfer from the conduction band of crTiO(2) to the nZVI on the crTiO(2) for the degradation of MO.
Sigma-Aldrich
Methyl Orange, SAJ first grade
Sigma-Aldrich
Methyl Orange, JIS special grade
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Methyl Orange
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4
Sigma-Aldrich
Methyl Orange solution, pH 3.1-4.4