Merck
  • A kinetic method for expeditious detection of pyrophosphate anions at nanomolar concentrations based on a nucleic acid fluorescent sensor.
A kinetic method for expeditious detection of pyrophosphate anions at nanomolar concentrations based on a nucleic acid fluorescent sensor.
Chemical communications (Cambridge, England) (2012-12-12)
Xin Su, Chen Zhang, Xianjin Xiao, Anqin Xu, Zhendong Xu, Meiping Zhao
ABSTRACT
A kinetic method has been developed for highly sensitive and selective detection of pyrophosphate anions (PPi) by using a fluorophore labelled single-stranded-DNA-Al(III) complex system. The facile and spectrum-tunable fluorescent sensor enables rapid detection of PPi in urine and cell lysate solutions without the need for complex pre-cleanup procedures.
Sigma-Aldrich
Pyrophosphoric acid, SAJ first grade, ≥90.0%
Sigma-Aldrich
Pyrophosphoric acid, technical grade