Merck
  • Quercetin supplementation is effective in improving mitochondrial dysfunctions induced by 3-nitropropionic acid: implications in Huntington's disease.
Quercetin supplementation is effective in improving mitochondrial dysfunctions induced by 3-nitropropionic acid: implications in Huntington's disease.
Biochimica et biophysica acta (2012-12-12)
Rajat Sandhir, Arpit Mehrotra
ABSTRACT
The study was designed to investigate the beneficial effect of quercetin supplementation in 3-nitropropionic acid (3-NP) induced model of Huntington's disease (HD). HD was induced in rats by administering sub-chronic dose of 3-NP, intraperitoneally, twice daily for 17days. Quercetin was supplemented at a dose of 25mg/kg body weight by oral gavage for 21days. At the end of treatment, mitochondrial bioenergetics, mitochondrial swelling, oxidative stress, neurobehavioral deficits and histopathological changes were analyzed. Quercetin supplementation was able to reverse 3-NP induced inhibition of respiratory chain complexes, restore ATP levels, attenuate mitochondrial oxidative stress in terms of lipid peroxidation and prevent mitochondrial swelling. Quercetin administration also restored the activities of superoxide dismutase and catalase along with thiol content in 3-NP treated animals. Beneficial effect of quercetin administration was observed on 3-NP induced motor deficits analyzed by narrow beam walk and footprint analysis. Histopathological analysis of 3-NP treated rats revealed pyknotic nuclei and astrogliosis in striatum, which were reduced or absent in quercetin supplemented animals. Altogether, our results show that quercetin supplementation to 3-NP induced HD animals ameliorated mitochondrial dysfunctions, oxidative stress and neurobehavioral deficits in rats showing potential of this flavonoid in maintaining mitochondrial functions, suggesting a putative role of quercetin in HD management.
Sigma-Aldrich
3-Nitropropionic acid, ≥97%