Merck
  • Single-solute and bisolute sorption of phenol and trichloroethylene from aqueous solution onto modified montmorillonite and application of sorption models.
Single-solute and bisolute sorption of phenol and trichloroethylene from aqueous solution onto modified montmorillonite and application of sorption models.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-11-07)
C D Wu, L Wang, C X Hu, M H He
ABSTRACT
The single-solute and bisolute sorption behaviour of phenol and trichloroethylene, two organic compounds with different structures, onto cetyltrimethylammonium bromide (CTAB)-montmorillonite was studied. The monolayer Langmuir model (MLM) and empirical Freundlich model (EFM) were applied to the single-solute sorption of phenol or trichloroethylene from water onto monolayer or multilayer CTAB-montmorillonite. The parameters contained in the MLM and EFM were determined for each solute by fitting to the single-solute isotherm data, and subsequently utilized in binary sorption. The extended Langmuir model (ELM) coupled with the single-solute MLM and the ideal adsorbed solution theory (IAST) coupled with the single-solute EFM were used to predict the binary sorption of phenol and trichloroethylene onto CTAB-montmorillonite. It was found that the EFM was better than the MLM at describing single-solute sorption from water onto CTAB-montmorillonite, and the IAST was better than the ELM at describing the binary sorption from water onto CTAB-montmorillonite.
Sigma-Aldrich
Nanoclay, hydrophilic bentonite
Sigma-Aldrich
Bentonite, CP
Sigma-Aldrich
Bentonite