Merck
  • Selective nanodecoration of modified cyclodextrin crystals with gold nanorods.
Selective nanodecoration of modified cyclodextrin crystals with gold nanorods.
Journal of colloid and interface science (2012-10-16)
Bárbara Herrera, Carolina Adura, Nicolás Yutronic, Marcelo J Kogan, Paul Jara
ABSTRACT
Gold nanorods (AuNRs) stabilized by cetyltrimethylammonium bromide (CTAB) were deposited onto crystals of α-cyclodextrin (α-CD) inclusion compounds (ICs) that contained octanethiol (OT) as guest molecules. The nanodecoration was produced specifically at the {001} crystal planes through interaction between the -SH groups of the ICs and the AuNRs.
Sigma-Aldrich
α-Cyclodextrin, ≥98%
Sigma-Aldrich
α-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
α-Cyclodextrin, purum, ≥98.0% (HPLC)
Sigma-Aldrich
α-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.0-101.0% cyclodextrin basis (HPLC)
Sigma-Aldrich
α-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (HPLC)
Sigma-Aldrich
1-Octanethiol, ≥98.5%