Merck
  • Electrochemical oxidation of pyrogallol: formation and characterization of long-lived oxygen radicals and application to assess the radical scavenging abilities of antioxidants.
Electrochemical oxidation of pyrogallol: formation and characterization of long-lived oxygen radicals and application to assess the radical scavenging abilities of antioxidants.
The journal of physical chemistry. B (2012-09-27)
Shaolin Mu, Chong Chen
ABSTRACT
Electrochemical oxidation of pyrogallol in a pH 5.0 phosphate buffer was carried out on a reduced graphene oxide/glassy carbon (RGO/GC) electrode. Reduced graphene oxide plays an important role in catalyzing the electrochemical oxidation of pyrogallol. A deep yellow film deposited on the electrode exhibits electroactivity in a wide pH range. On the basis of the experimental results from measurements of (1)H NMR, (13)C NMR, and IR spectra, there are hydroxyl, carbonyl, and aldehyde groups in the product. No visible absorption peak occurs in the UV-vis spectrum of the product, and its molecular weight is lower than that of the dipolymer but higher than that of the monomer. Therefore, the film is neither a polymer nor a dipolymer and is only a product of pyrogallol oxidation with oxygen radicals. No tendency toward the decay of the ESR signal intensity of the electrogenerated film deposited on the RGO/graphite electrode was observed after 210 days. Electrogenerated film was used as a radical source to test the radical scavenging abilities of ascorbic acid, catechin, and catechol in aqueous solutions based on the ESR signal intensity. The result indicates that ascorbic acid and catechin can scavenge the free radicals, but catechol can hardly scavenge the free radicals.
Product Description
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Pyrogallol, JIS special grade, ≥99.0%