Merck
  • Unprecedented hydroxyl radical-dependent two-step chemiluminescence production by polyhalogenated quinoid carcinogens and H2O2.
Unprecedented hydroxyl radical-dependent two-step chemiluminescence production by polyhalogenated quinoid carcinogens and H2O2.
Proceedings of the National Academy of Sciences of the United States of America (2012-09-19)
Ben-Zhan Zhu, Li Mao, Chun-Hua Huang, Hao Qin, Rui-Mei Fan, Balaraman Kalyanaraman, Jun-Ge Zhu
ABSTRACT
Most chemiluminescence (CL) reactions usually generate only one-step CL, which is rarely dependent on the highly reactive and biologically/environmentally important hydroxyl radicals ((•)OH). Here, we show that an unprecedented two-step CL can be produced by the carcinogenic tetrachloro-1,4-benzoquinone (also known as p-chloranil) and H(2)O(2), which was found to be well-correlated to and directly dependent on its two-step metal-independent production of (•)OH. We proposed that (•)OH-dependent formation of quinone-dioxetane and electronically excited carbonyl species might be responsible for this unusual two-step CL production by tetrachloro-1,4-benzoquinone/H(2)O(2). This is a unique report of a previously undefined two-step CL-producing system that is dependent on intrinsically formed (•)OH. These findings may have potential applications in detecting and quantifying (•)OH and the ubiquitous polyhalogenated aromatic carcinogens, which may have broad biological and environmental implications for future research on these types of important species.
Sigma-Aldrich
Tetrachloro-1,4-benzoquinone, 99%