Merck
  • Effects of alkyl chain length, solvent and tandem Claisen rearrangement on two-dimensional structures of noncyclic isobutenyl compounds: scanning tunnelling microscopic study.
Effects of alkyl chain length, solvent and tandem Claisen rearrangement on two-dimensional structures of noncyclic isobutenyl compounds: scanning tunnelling microscopic study.
Organic & biomolecular chemistry (2012-09-12)
Yoshihiro Kikkawa, Kazuhiro Omori, Mayuko Takahashi, Masatoshi Kanesato, Kazuhisa Hiratani
ABSTRACT
A series of isobutenyl compounds possessing various alkyl chain lengths (C(n)-1) with a carbon number of n = 14-21 were synthesized and their two-dimensional (2D) structures were systematically studied using scanning tunnelling microscopy (STM) at a highly oriented pyrolytic graphite (HOPG)/solvent interface. Two kinds of solvent, such as 1-phenyloctane (PO) and 1-phenylnonane (PN), were selected to examine the 2D structures by changing the alkyl chain length of the isobutenyl compounds. At the HOPG/PO interface, C(n)-1 molecules with shorter alkyl chains (n = 14-17) showed the same zig-zag shaped 2D structure regardless of the alkyl chain length, whereas an odd-even effect was recognized in C(n)-1 compounds with longer alkyl chains (n = 18-21) displaying the wavy and tripod structures, alternately. This odd-even effect was also observed at the HOPG/PN interface rather more distinctly. These results suggest that there is a specific alkyl chain length range that shows the odd-even effect in the present 2D system. After a tandem Claisen rearrangement (TCR), the 2D structures of all the C(n)-2 compounds formed were converged into the same linear structure, i.e. the odd-even effect was cancelled by the conformational limitation induced by the TCR.
Sigma-Aldrich
1-Phenylnonane, 96%
Sigma-Aldrich
2-Methylpropene, 99%