Merck
  • Aloe barbadensis Miller mediated green synthesis of mono-disperse copper oxide nanoparticles: optical properties.
Aloe barbadensis Miller mediated green synthesis of mono-disperse copper oxide nanoparticles: optical properties.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-09-04)
Sangeetha Gunalan, Rajeshwari Sivaraj, Rajendran Venckatesh
ABSTRACT
In this paper, we report on the synthesis of nanostructured copper oxide particles by both chemical and biological method. A facile and efficient synthesis of copper oxide nanoparticles was carried out with controlled surface properties via green chemistry approach. The CuO nanoparticles synthesized are monodisperse and versatile and were characterized with the help of UV-Vis, PL, FT-IR, XRD, SEM, and TEM techniques. The particles are crystalline in nature and average sizes were between 15 and 30 nm. The morphology of the nanoparticles can be controlled by tuning the amount of Aloe vera extract. This new eco-friendly approach of synthesis is a novel, cheap, and convenient technique suitable for large scale commercial production and health related applications of CuO nanoparticles.
Sigma-Aldrich
Copper(I) oxide, ≥99.99% trace metals basis, anhydrous
Sigma-Aldrich
Copper(I) oxide, powder, ≤7 μm, 97%