Merck
Study of the total synthesis of (-)-exiguolide.
The Journal of organic chemistry (2012-08-02)
Cyril Cook, Frédéric Liron, Xavier Guinchard, Emmanuel Roulland
ABSTRACT
In this article, we disclose the various routes and strategies we had to explore before finally achieving the total synthesis of (-)-exiguolide ((-)-1). Two first types of approaches were set, both relying on the Trost's domino ene-yne coupling/oxa-Michael reaction that we choose for its ability to control the geometry of the methylacrylate-bearing tetrahydropyrane ring B. In our first approach, we expected to assemble the two main fragments (C14-C21 and C1-C13) by creating the C13-C14 bond through a palladium(0)-catalyzed cross-coupling, but this step failed, unfortunately. In the second approach, which was more linear, we created the C16-C17 bond through condensation of a lithium acetylide on a Weinreb amide, and we assembled the C1-C5 and C6-C21 subunits through Trost's domino ene-yne coupling/oxa-Michael reaction. These two approaches served us to design an ameliorated third strategy, which finally led to the total synthesis of (-)-exiguolide.
Product Description
Supelco
Methyl acrylate, analytical standard
Sigma-Aldrich
Methyl acrylate, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor