Merck
  • Characterization of the nitrobenzene-degrading strain Pseudomonas sp. a3 and use of its immobilized cells in the treatment of mixed aromatics wastewater.
Characterization of the nitrobenzene-degrading strain Pseudomonas sp. a3 and use of its immobilized cells in the treatment of mixed aromatics wastewater.
World journal of microbiology & biotechnology (2012-07-19)
Zhiguo Wu, Yalong Liu, Hongming Liu, Yali Xia, Wenjing Shen, Qing Hong, Shunpeng Li, Hangyong Yao
ABSTRACT
A bacterial strain Pseudomonas sp. a3 capable of degrading nitrobenzene, phenol, aniline, and other aromatics was isolated and characterized. When nitrobenzene was degraded, the release of NH(4) (+) was detected, but not of NO(2) (-). This result implied that nitrobenzene might have a partial reductive metabolic pathway in strain a3. However, aniline appeared as one of the metabolites during the aerobic degradation of nitrobenzene. Moreover, the appearance of 2-aminophenol during aniline degradation by strain a3 indicated that novel initial reactions existed during the degradation of nitrobenzene and aniline by strain a3. Strain a3 was immobilized in the mixed carrier of polyvinyl alcohol and sodium alginate to improve its degrading efficiency. The optimal concentrations of polyvinyl alcohol and sodium alginate in the mixed carrier were 9 and 3 %, respectively. The immobilized cells had stable degradation activity and good mechanical properties in the recycling tests. The immobilized cells also exhibited higher tolerances in acidic (pH 4-5) and highly saline (10 % NaCl) environments than those of free cells. The biodegradation of nitrobenzene mixed with aniline and phenol using immobilized cells of Pseudomonas sp. a3 was also greatly improved compared with those of free cells. The immobilized cells could completely degrade 300 mg L(-1) nitrobenzene within 10 h with 150 mg L(-1) aniline and 150 mg L(-1) phenol. This result revealed that the immobilized cells of Pseudomonas sp. a3 could be a potential candidate for treating nitrobenzene wastewater mixed with other aromatics.
Product Description
Supelco
Nitrobenzene, PESTANAL®, analytical standard
Sigma-Aldrich
Nitrobenzene, ReagentPlus®, 99%
Sigma-Aldrich
Nitrobenzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Nitrobenzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Nitrobenzene, ACS reagent, ≥99.0%