Merck
Total synthesis and antifungal activity of (2S,3R)-2-aminododecan-3-ol.
Bioorganic & medicinal chemistry letters (2012-06-19)
T Vijai Kumar Reddy, B L A Prabhavathi Devi, R B N Prasad, M Poornima, C Ganesh Kumar
ABSTRACT
We report the total synthesis of (2S,3R)-2-aminododecan-3-ol has been achieved starting from commercially available 10-undecenoic acid. The key steps involved are Sharpless asymmetric epoxidation, Miyashita's boron-directed C-2 regioselective azidolysis, generated the asymmetric centers and in situ detosylation and reduction of azido tosylate. The antifungal activity of the synthesized (2S,3R)-2-aminododecan-3-ol was evaluated on several Candida strains and was comparable to miconazole, a standard drug.
Product Description
Supelco
1-Dodecanol, Selectophore, ≥98.0%
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
Sigma-Aldrich
1-Dodecanol, ACS reagent, ≥98.0%
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%