Merck
  • Selective production of hybridoma cells: antigenic-based pre-selection of B lymphocytes for electrofusion with myeloma cells.
Selective production of hybridoma cells: antigenic-based pre-selection of B lymphocytes for electrofusion with myeloma cells.
Biochimica et biophysica acta (1990-12-10)
M Tomita, T Y Tsong
ABSTRACT
Methods for pre-selecting B lymphocytes were studied and investigated. First, biotinylated antigen was used for selecting B lymphocytes. These pre-selected B lymphocytes were then combined with biotinylated myeloma cells by adding streptavidin. The final formula of the selected B cell-myeloma cell was as follows: B cell-(antigen-biotin-strept-avidin-biotin)-myeloma cell. Then, this B cell-myeloma cell conjugate was fused by the pulsed electric field (PEF) method, which fused only those conjugated cells. The fusion efficiency obtained by this method was 3-15-times higher than that obtained by the non-specific poly(ethylene glycol) (PEG) fusion method. Second, avidin-antigen conjugate was used to select B lymphocytes. For this purpose, bifunctional cross-linkers such as N-succinimidyl 3-(2-pyridyldithio) propionate (SPDP) and m-maleimidobenzoyl N-hydroxysuccinimide (MBS) were chosen. Each reagent contains two heterofunctional groups which can make covalent bond with both Lys and Cys residues. Typical avidin-antigen conjugate is expressed as avidin-SPDP (or MBS)-antigen. Thus, final B cell-myeloma cell conjugate was B cell-antigen-SPDP (or MBS)-avidin-biotin-myeloma cell. The yield of this procedure was of the order of 10(-2). Here, we suggest that the pre-selection of B lymphocytes by biotinylated antigen or avidin-antigen conjugate is a new method of obtaining selected hybridoma cells which produce specific monoclonal antibodies against the antigen used for selecting B lymphocytes.