Merck
  • Organocatalyzed enantioselective formal [4 + 2] cycloaddition of 2,3-disubstituted indole and methyl vinyl ketone.
Organocatalyzed enantioselective formal [4 + 2] cycloaddition of 2,3-disubstituted indole and methyl vinyl ketone.
Organic letters (2012-05-26)
Quan Cai, Shu-Li You
ABSTRACT
A formal [4 + 2] cycloaddition of 2,3-disubstituted indoles with vinyl methyl ketone was realized in the presence of a catalytic amount of quinine-derived primary amine and pentafluorobenzoic acid. This method provides bridged-ring indoline scaffolds containing two quaternary carbon centers with excellent yields and enantioselectivity (up to 98% yield and 98% ee).
Sigma-Aldrich
3-Buten-2-one, contains 0.3-1.0% hydroquinone as stabilizer, technical grade, 90%