Merck
  • Plasticity of spatial hearing: behavioural effects of cortical inactivation.
Plasticity of spatial hearing: behavioural effects of cortical inactivation.
The Journal of physiology (2012-05-02)
Fernando R Nodal, Victoria M Bajo, Andrew J King
ABSTRACT
The cerebral cortex plays a critical role in perception and in learning-induced plasticity. We show that reversibly silencing any of the main regions of auditory cortex impairs the ability of adult ferrets to localize sound, with the largest deficit seen after deactivating the primary fields. Although these animals had no trouble localizing longer sound bursts, their performance dropped considerably when auditory spatial cues were altered by occluding one ear with an earplug. In contrast to control ferrets, which recovered their localization abilities with intensive training, adaptation to an earplug was impaired following cortical inactivation, with the greatest disruption in plasticity observed after silencing higher-level cortical areas. These findings imply regional differences in the processing of spatial information across the auditory cortex.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)