Merck
  • Continuous and segmented flow microfluidics: applications in high-throughput chemistry and biology.
Continuous and segmented flow microfluidics: applications in high-throughput chemistry and biology.
Chimia (2012-05-02)
Claire E Stanley, Robert C R Wootton, Andrew J deMello
ABSTRACT
This account highlights some of our recent activities focused on developing microfluidic technologies for application in high-throughput and high-information content chemical and biological analysis. Specifically, we discuss the use of continuous and segmented flow microfluidics for artificial membrane formation, the analysis of single cells and organisms, nanomaterial synthesis and DNA amplification via the polymerase chain reaction. In addition, we report on recent developments in small-volume detection technology that allow access to the vast amounts of chemical and biological information afforded by microfluidic systems.
Sigma-Aldrich
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines
Sigma-Aldrich
Fluorescamine, ≥98.0%