Merck
  • Detection of neural activity in the brains of Japanese honeybee workers during the formation of a "hot defensive bee ball".
Detection of neural activity in the brains of Japanese honeybee workers during the formation of a "hot defensive bee ball".
PloS one (2012-03-21)
Atsushi Ugajin, Taketoshi Kiya, Takekazu Kunieda, Masato Ono, Tadaharu Yoshida, Takeo Kubo
ABSTRACT
Anti-predator behaviors are essential to survival for most animals. The neural bases of such behaviors, however, remain largely unknown. Although honeybees commonly use their stingers to counterattack predators, the Japanese honeybee (Apis cerana japonica) uses a different strategy to fight against the giant hornet (Vespa mandarinia japonica). Instead of stinging the hornet, Japanese honeybees form a "hot defensive bee ball" by surrounding the hornet en masse, killing it with heat. The European honeybee (A. mellifera ligustica), on the other hand, does not exhibit this behavior, and their colonies are often destroyed by a hornet attack. In the present study, we attempted to analyze the neural basis of this behavior by mapping the active brain regions of Japanese honeybee workers during the formation of a hot defensive bee ball. First, we identified an A. cerana homolog (Acks = Apis cerana kakusei) of kakusei, an immediate early gene that we previously identified from A. mellifera, and showed that Acks has characteristics similar to kakusei and can be used to visualize active brain regions in A. cerana. Using Acks as a neural activity marker, we demonstrated that neural activity in the mushroom bodies, especially in Class II Kenyon cells, one subtype of mushroom body intrinsic neurons, and a restricted area between the dorsal lobes and the optic lobes was increased in the brains of Japanese honeybee workers involved in the formation of a hot defensive bee ball. In addition, workers exposed to 46°C heat also exhibited Acks expression patterns similar to those observed in the brains of workers involved in the formation of a hot defensive bee ball, suggesting that the neural activity observed in the brains of workers involved in the hot defensive bee ball mainly reflects thermal stimuli processing.
Sigma-Aldrich
Isoamyl acetate, natural, ≥97%, FCC, FG
Sigma-Aldrich
Isoamyl acetate, ≥95%, FCC, FG
Sigma-Aldrich
Isopentyl acetate, SAJ special grade, ≥98.0%
Sigma-Aldrich
Isopentyl acetate, mixture of isomers, reagent grade, 98%
Supelco
Isopentyl acetate, analytical standard
Sigma-Aldrich
Isopentyl acetate, anhydrous, ≥99%