Merck
  • Nitric oxide production mediates oligogalacturonide-triggered immunity and resistance to Botrytis cinerea in Arabidopsis thaliana.
Nitric oxide production mediates oligogalacturonide-triggered immunity and resistance to Botrytis cinerea in Arabidopsis thaliana.
Plant, cell & environment (2012-03-08)
S Rasul, C Dubreuil-Maurizi, O Lamotte, E Koen, B Poinssot, G Alcaraz, D Wendehenne, S Jeandroz
ABSTRACT
Nitric oxide (NO) regulates a wide range of plant processes from development to environmental adaptation. In this study, we investigated the production and/or function of NO in Arabidopsis thaliana leaf discs and plants elicited by oligogalacturonides (OGs) and challenged with Botrytis cinerea. We provided evidence that OGs triggered a fast and long lasting NO production which was Ca(2+) dependent and involved nitrate reductase (NR). Accordingly, OGs triggered an increase of both NR activity and transcript accumulation. NO production was also sensitive to the mammalian NO synthase inhibitor L-NAME. Intriguingly, we showed that L-NAME affected NO production by interfering with NR activity, thus questioning the mechanisms of how this compound impairs NO synthesis in plants. We further demonstrated that NO modulates RBOHD-mediated reactive oxygen species (ROS) production and participates in the regulation of OG-responsive genes such as anionic peroxidase (PER4) and a β-1,3-glucanase. Mutant plants impaired in PER4 and β-1,3-glucanase, as well as Col-0 plants treated with the NO scavenger cPTIO, were more susceptible to B. cinerea. Taken together, our investigation deciphers part of the mechanisms linking NO production, NO-induced effects and basal resistance to B. cinerea.
Sigma-Aldrich
D-Galacturonic acid sodium salt, ≥95.0% (T)