Merck
  • Effects of a wheat bran extract containing arabinoxylan oligosaccharides on gastrointestinal health parameters in healthy adult human volunteers: a double-blind, randomised, placebo-controlled, cross-over trial.
Effects of a wheat bran extract containing arabinoxylan oligosaccharides on gastrointestinal health parameters in healthy adult human volunteers: a double-blind, randomised, placebo-controlled, cross-over trial.
The British journal of nutrition (2012-03-01)
Isabelle E J A François, Olivier Lescroart, Wim S Veraverbeke, Massimo Marzorati, Sam Possemiers, Pieter Evenepoel, Henrike Hamer, Els Houben, Karen Windey, Gjalt W Welling, Jan A Delcour, Christophe M Courtin, Kristin Verbeke, Willem F Broekaert
ABSTRACT
Wheat bran extract (WBE) is a food-grade soluble fibre preparation that is highly enriched in arabinoxylan oligosaccharides. In this placebo-controlled cross-over human intervention trial, tolerance and effects on colonic protein and carbohydrate fermentation were studied. After a 1-week run-in period, sixty-three healthy adult volunteers consumed 3, 10 and 0 g WBE/d for 3 weeks in a random order, with 2 weeks' washout between each treatment period. Fasting blood samples were collected at the end of the run-in period and at the end of each treatment period for analysis of haematological and clinical chemistry parameters. Additionally, subjects collected a stool sample for analysis of microbiota, SCFA and pH. A urine sample, collected over 48 h, was used for analysis of p-cresol and phenol content. Finally, the subjects completed questionnaires scoring occurrence frequency and distress severity of eighteen gastrointestinal symptoms. Urinary p-cresol excretion was significantly decreased after WBE consumption at 10 g/d. Faecal bifidobacteria levels were significantly increased after daily intake of 10 g WBE. Additionally, WBE intake at 10 g/d increased faecal SCFA concentrations and lowered faecal pH, indicating increased colonic fermentation of WBE into desired metabolites. At 10 g/d, WBE caused a mild increase in flatulence occurrence frequency and distress severity and a tendency for a mild decrease in constipation occurrence frequency. In conclusion, WBE is well tolerated at doses up to 10 g/d in healthy adults volunteers. Intake of 10 g WBE/d exerts beneficial effects on gut health parameters.
Product Description
Supelco
4-Methylphenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
p-Cresol, JIS special grade, ≥99.0%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
p-Cresol, 99%
Supelco
p-Cresol, analytical standard