Merck
  • Tropicibacter phthalicus sp. nov., a phthalate-degrading bacterium from seawater.
Tropicibacter phthalicus sp. nov., a phthalate-degrading bacterium from seawater.
Current microbiology (2012-01-27)
Hiroaki Iwaki, Ayaka Nishimura, Yoshie Hasegawa
ABSTRACT
An aerobic, Gram-negative bacterial strain, designated KU27E1(T), which degrades phthalate and dimethylphthalate, was isolated from seawater obtained from the coastal region of Ishigaki Island, Japan. Cells are motile rods with polar flagella. Strain KU27E1(T) grew at 15-30°C, pH 6.0-8.0, in the presence of 1.0-2.0% (w/v) NaCl. The 16S rRNA gene sequence analysis revealed that this strain was affiliated with the family Rhodobacteraceae in the class Alphaproteobacteria, and was most closely related to Tropicibacter naphthalenivorans (96.8%). The predominant respiratory lipoquinone was ubiquinone-10, and the major cellular fatty acid was C(18:1)ω7c (88.5%). The G+C content of genomic DNA was 58.7 mol%. Based on the physiological, chemotaxonomic, and phylogenetic data, strain KU27E1(T) is suggested to represent a novel species of the genus Tropicibacter, for which the name Tropicibacter phthalicus sp. nov. is proposed. The type strain of Tropicibacter phthalicus is designated as KU27E1(T) (=JCM 17793(T) = KCTC 23703(T)).
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Sigma-Aldrich
Dimethyl phthalate, ≥99%
Supelco
Dimethyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dimethyl phthalate, SAJ special grade, ≥99.0%