Merck
  • Development of cloxacillin loaded multiple-unit alginate-based floating system by emulsion-gelation method.
Development of cloxacillin loaded multiple-unit alginate-based floating system by emulsion-gelation method.
International journal of biological macromolecules (2011-10-25)
Jadupati Malakar, Amit Kumar Nayak, Dilipkumar Pal
ABSTRACT
This work investigates the development, optimization and in vitro evaluation of liquid paraffin-entrapped multiple-unit alginate-based floating system containing cloxacillin by emulsion-gelation method for gastro retentive delivery. The effect of process variables like drug to polymer ratio by weight, and liquid paraffin to water ratio by volume on various physicochemical properties in case of liquid paraffin-entrapped calcium alginate beads containing cloxacillin applicable to drug entrapment efficiency, density and drug release was optimized using 3(2) factorial design and analyzed using response surface methodology. The observed (actual values) responses were coincided well with the predicted values, given by the optimization technique. The optimized beads showed drug entrapment efficiency of 64.63±0.78%, density of 0.90±0.05 g/cm(3), and drug release of 56.72±0.85% in simulated gastric fluid (pH 1.2) after 8h with floating lag time of 8.45 min and floated well over 12h in simulated gastric fluid (pH 1.2). The average size of all dried beads ranged from 1.73±0.04 to 1.97±0.08 mm. The beads were characterized by SEM and FTIR for surface morphology and excipients-drug interaction analysis, respectively. All these beads showed prolonged sustained release of cloxacillin over 8h in simulated gastric fluid (pH 1.2). The cloxacillin release profile from liquid paraffin beads followed Korsmeyer-Peppas model over a period of 8h with anomalous (non-Fickian) diffusion mechanism for drug release.
Sigma-Aldrich
Cloxacillin sodium salt monohydrate, β-lactamase-resistant antibiotic