Merck
  • Integration of diverse data sources for prediction of adverse drug events.
Integration of diverse data sources for prediction of adverse drug events.
Clinical pharmacology and therapeutics (2011-10-21)
D R Abernethy, J P F Bai, K Burkhart, H-G Xie, P Zhichkin
ABSTRACT
The rapid evolution of large biological, pharmacological, and chemical databases has led to optimism that such data resources can be leveraged for prediction of drug action based on molecular descriptors of the drug. Challenges to realize this possibility include organization of each type of database in a manner that allows extraction of information across disparate data sources and the linkage of information across the biological, pharmacological, and chemical domains.
Product Description
Supelco
Cyclobenzaprine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Cyclobenzaprine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cyclobenzaprine hydrochloride