Merck
  • Development of an electrochemical Limulus amebocyte lysate assay technique for portable and highly sensitive endotoxin sensor.
Development of an electrochemical Limulus amebocyte lysate assay technique for portable and highly sensitive endotoxin sensor.
Innate immunity (2011-08-17)
Kumi Y Inoue, Satoko Takahashi, Kosuke Ino, Hitoshi Shiku, Tomokazu Matsue
ABSTRACT
Here, we report the development of an electrochemical detection method for endotoxin based on the Limulus amebocyte lysate (LAL) assay. A mixture of LAL reagent and endotoxin sample solution was incubated for 1 h. The endotoxin activated a cascade reaction of zymogens contained in the LAL to generate p-nitroaniline (pNA) which was then electrochemically detected by differential pulse voltammetry (DPV). The generated pNA gave a clear peak at -0.75 V vs. silver/silver chloride (Ag/AgCl), which increased with the concentration of endotoxin in the LAL assay solution. This DPV detection was performed using an electrode chip device fabricated from a diamond-like carbon-coated glass substrate. This chip device could detect as low as 10 endotoxin units l(-1) at room temperature within 1 h. This novel electrochemical method for the detection of endotoxin appears promising for the development of compact, low-cost and easy-to-use sensors for on-site monitoring of potentially contaminated medical supplies, including dialysis fluid, transplanted tissue and culture medium for assisted reproduction.
Sigma-Aldrich
4-Nitroaniline, ≥99%
Supelco
4-Nitroaniline, analytical standard