Merck
  • Role of angiotensin II in plasma PAI-1 changes induced by imidapril or candesartan in hypertensive patients with metabolic syndrome.
Role of angiotensin II in plasma PAI-1 changes induced by imidapril or candesartan in hypertensive patients with metabolic syndrome.
Hypertension research : official journal of the Japanese Society of Hypertension (2011-08-05)
Roberto Fogari, Annalisa Zoppi, Amedeo Mugellini, Pamela Maffioli, Pierangelo Lazzari, Giuseppe Derosa
ABSTRACT
To evaluate the relationship between plasma plasminogen activator inhibitor-1 (PAI-1) and angiotensin II (Ang II) changes during treatment with imidapril and candesartan in hypertensive patients with metabolic syndrome. A total of 84 hypertensive patients with metabolic syndrome were randomized to imidapril 10 mg or candesartan 16 mg for 16 weeks. At weeks 4 and 8, there was a dose titration to imidapril 20 mg and candesartan 32 mg in nonresponders (systolic blood pressure (SBP) >140 and/or diastolic blood pressure (DBP) >90 mm Hg). We evaluated, at baseline and after 2, 4, 8, 12 and 16 weeks, clinic blood pressure, Ang II and PAI-1 antigen. Both imidapril and candesartan induced a similar SBP/DBP reduction (-19.4/16.8 and -19.5/16.3 mm Hg, respectively, P<0.001 vs. baseline). Both drugs decreased PAI-1 antigen after 4 weeks of treatment, but only the PAI-1 lowering effect of imidapril was sustained throughout the 16 weeks (-9.3 ng ml(-1), P<0.01 vs. baseline), whereas candesartan increased PAI-1 (+6.5 ng ml(-1), P<0.05 vs. baseline and P<0.01 vs. imidapril). Imidapril significantly decreased Ang II levels (-14.6 pg ml(-1) at week 16, P<0.05 vs. baseline), whereas candesartan increased them (+24.2 pg ml(-1), P<0.01 vs. baseline and vs. imidapril). In both groups there was a positive correlation between Ang II and PAI-1 changes (r=0.61, P<0.001 at week 16 for imidapril, and r=0.37, P<0.005 at week 16 for candesartan). Imidapril reduced plasma PAI-1 and Ang II levels, whereas candesartan increased them. This suggests that the different effect of angiotensin-converting enzyme inhibitors and Ang II blockers on Ang II production has a role in their different influence on fibrinolysis.
Sigma-Aldrich
Imidapril hydrochloride, ≥98% (HPLC)