Merck
  • Stereocontrolled synthesis of α-amino-α'-alkoxy ketones by a copper-catalyzed cross-coupling of peptidic thiol esters and α-alkoxyalkylstannanes.
Stereocontrolled synthesis of α-amino-α'-alkoxy ketones by a copper-catalyzed cross-coupling of peptidic thiol esters and α-alkoxyalkylstannanes.
Organic letters (2011-06-17)
Hao Li, Anyu He, John R Falck, Lanny S Liebeskind
ABSTRACT
A stereocontrolled synthesis of α-amino-α'-alkoxy ketones is described. This pH-neutral copper(I) thiophene-2-carboxylate (CuTC)-catalyzed cross-coupling of amino acid thiol esters and chiral nonracemic α-alkoxyalkylstannanes gives α-amino-α'-alkoxy ketones in good to excellent yields with complete retention of configuration at the α-amino- and α-alkoxy-substituted stereocenters.
Sigma-Aldrich
2-Thiophenecarboxylic acid, ReagentPlus®, 99%