Merck
  • Structure of an exocellular polysaccharide produced by Streptococcus thermophilus.
Structure of an exocellular polysaccharide produced by Streptococcus thermophilus.
Carbohydrate research (1990-05-01)
T Doco, J M Wieruszeski, B Fournet, D Carcano, P Ramos, A Loones
ABSTRACT
Streptococcus thermophilus strains grown on skimmed milk produced a viscosifying, exocellular, and water-soluble polysaccharide which contains D-glucose, D-galactose, and N-acetyl-D-galactosamine in the ratio of 1:2:1. Methylation analysis identified the glycosidic linkages in the tetrasaccharidic repeating-unit, and Smith degradation and nitrous deamination after N-deacetylation gave the sequence of monosaccharides in the repeating-unit. The anomeric configurations of the sugar residues were determined by oxidation of the peracetylated polysaccharide with chromium trioxide and by 1H- and 13C-n.m.r. spectroscopy. The following structure was assigned to the repeating unit of the polysaccharide,----3)-beta-D-Galp-(1----3)-[alpha-D-Galp-(1----6)]-beta- D- Glcp-(1----3)-alpha-D-GalpNAc-(1----.
Sigma-Aldrich
Chromium(VI) oxide, SAJ special grade, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, 99.99% trace metals basis
Sigma-Aldrich
Chromium(VI) oxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, ReagentPlus®, 99.9% trace metals basis