Merck
  • Determination of elaidic and vaccenic acids in foods using GC×GC-FID and GC×GC-TOFMS.
Determination of elaidic and vaccenic acids in foods using GC×GC-FID and GC×GC-TOFMS.
Talanta (2011-06-08)
Jaeho Ha, Dongwon Seo, Dongbin Shin
ABSTRACT
Trans fatty acids (TFAs) are present in meat and dairy products as m ruminant animals and in vegetable fats due to partial hydrogenation. This study aimed to discriminate between natural (N-TFA) and hydrogenated trans fatty (H-TFA) acids by GC×GC-flame ionization detection (GC×GC-FID) and comprehensive GC×GC-time-of-flight mass spectrometry (GC×GC-TOFMS). The separation of two kinds of trans fats, vaccenic acid (18:1 trans-11) and elaidic acid (18:1 trans-9), was performed using GC×GC-FID and GC×GC-TOFMS. A 100 m×0.25 mm I.D.×0.2 μm (film thickness) SP-2560 (bis-cyanopropyl polysiloxane) fused capillary column (first separation dimension, 1D) was coupled to a 1.5 m×0.18 mm I.D.×0.18 μm (film thickness) RTX-5 (5% diphenyl/95% dimethyl polysiloxane) fused capillary column (second separation dimension, 2D). The RSD of the intra-day repeatability by both GC×GC-FID and GC×GC-TOFMS for elaidic and vaccenic acids was ≤9.56% and ≤9.97%, and the RSD of the inter-day repeatability was ≤8.49 and ≤9.06%, respectively. It was found that the V/E value (vaccenic acid to elaidic acid ratio) could be used to distinguish H-TFA from N-TFA and to evaluate the quality of the fatty foods.
Product Description
Supelco
SP®-2560 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2560 Capillary GC Column, L × I.D. 75 m × 0.18 mm, df 0.14 μm
Supelco
Elaidic acid, analytical standard
Sigma-Aldrich
Elaidic acid, ≥99.0% (GC)
Supelco
SP®-2560 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2560 Capillary GC Column, L × I.D. 200 m × 0.25 mm, df 0.20 μm
Sigma-Aldrich
trans-Vaccenic acid, ≥99% (capillary GC)