Merck
Electrical detection of single methylcytosines in a DNA oligomer.
Journal of the American Chemical Society (2011-05-13)
Makusu Tsutsui, Kazuki Matsubara, Takahito Ohshiro, Masayuki Furuhashi, Masateru Taniguchi, Tomoji Kawai
ABSTRACT
We report label-free electrical detections of chemically modified nucleobases in a DNA using a nucleotide-sized electrode gap. We found that methyl substitution contributes to increase the tunneling conductance of deoxycytidines, which was attributed to a shift of the highest occupied molecular orbital level closer to the electrode Fermi level by methylation. We also demonstrate statistical identifications of methylcytosines in an oligonucleotide by tunneling current. This result suggests a possible use of the transverse electron-transport method for a methylation level analysis.
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%