Merck
  • Effects of common dissolved anions on the reduction of para-chloronitrobenzene by zero-valent iron in groundwater.
Effects of common dissolved anions on the reduction of para-chloronitrobenzene by zero-valent iron in groundwater.
Water science and technology : a journal of the International Association on Water Pollution Research (2011-04-22)
C Le, J H Wu, S B Deng, P Li, X D Wang, N W Zhu, P X Wu
ABSTRACT
Batch tests were conducted to evaluate the influences of several common dissolved anions in groundwater on the reduction of para-chloronitrobenzene (p-CNB) by zero-valent iron (ZVI). The results showed that p-CNB reduction was enhanced by both Cl(-) and SO(4)(2-). HCO(3)(-) could either improve or inhibit p-CNB reduction, depending on whether the mixing speed was intense enough to rapidly eliminate Fe-carbonate complex deposited on ZVI surface. Above a concentration of 100 mg L(-1), NO(3)(-) increased the p-CNB reduction rate. The reduction rate by ClO(4)(-) decreased because the ClO(4)(-) competed with p-CNB for electrons. The p-CNB reduction was inhibited by PO(4)(3-), SiO(3)(2-) and humic acid, in the order humic acid < PO(4)(3-) < SiO(3)(2-), since these ions could form inner-sphere complexes on iron surface. The reaction even ceased when the ion concentrations were greater than 4, 0.5, and 30 mg L(-1), respectively. The results indicated that common dissolved anions in groundwater should be taken into account when ZVI is applied for contaminated groundwater remediation.
Sigma-Aldrich
1-Chloro-4-nitrobenzene, 99%