Merck
  • Oxidative stress augments the secretion of atrial natriuretic peptide in isolated rat atria.
Oxidative stress augments the secretion of atrial natriuretic peptide in isolated rat atria.
Peptides (2011-03-29)
Amin Shah, Shan Gao, Young-Bin Oh, Woo Hyun Park, Suhn Hee Kim
ABSTRACT
Reactive oxygen species (ROS) play a role in cardiovascular diseases such as hypertension and heart failure. The objective of the present study was to investigate the role of endogenous ROS in atrial hemodynamics and ANP secretion in isolated perfused beating rat atria. Pyrogallol (a generator of superoxide anion, 0.1, 1mM) or hydrogen peroxide (0.1, 1, 10, 30mM) was perfused into atria paced at 1.2Hz. Pyrogallol and hydrogen peroxide stimulated ANP secretion and concentration in a dose-dependent manner and dramatically decreased atrial contractility and translocation of extracellular fluid. The stimulatory effect of pyrogallol and hydrogen peroxide on ANP secretion was attenuated by the pretreatment with ascorbic acid (an antioxidant; 1mM) and cariporide (an inhibitor of the Na(+)/H(+) exchanger; 1μM) but negative inotropic effect was not changed. U120 (a MAPK(erk) pathway inhibitor; 10μM) attenuated the stimulatory effect of hydrogen peroxide on ANP secretion. However, U120 augmented negative inotropic effect and stimulatory effect of ANP concentration induced by pyrogallol. Antioxidant such as N-acetyl cystein, gallate, propyl gallate, or ellagic acid did not cause any significant changes in atrial parameters. These results suggest that intracellular - formed ROS stimulates ANP secretion partly through activation of MAPK(erk) pathway and Na(+)/H(+) exchanger.
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, powder
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material