Merck
  • [Effects of chlorimuron-ethyl on soil microbial community structure in soybean field].
[Effects of chlorimuron-ethyl on soil microbial community structure in soybean field].
Ying yong sheng tai xue bao = The journal of applied ecology (2011-03-03)
Yu Sheng, Jun Xu, Xin-gang Liu, Feng-shou Dong, Chang-peng Zhang, Cai-hong Lu, Yong-quan Zheng
ABSTRACT
By using phospholipid fatty acids (PLFA) method, this paper studied the soil microbial community structure in the soybean fields with different chlorimuron-ethyl application history in Weihe district of Heilongjiang Province. In the meantime, the residual amount of chlorimuron-ethyl in soil was determined. There was a very low residual of chlorimuron-ethyl in soil under the conditions of different chlorimuron-ethyl application history. With the increasing year of chlorimuron-ethyl application, the total concentration of soil microbial PLFA and the ratios of fungi/bacteria and Gram negative/Gram positive bacteria decreased, and the microbial stress level increased. Principal component analysis (PCA) showed that long-term application of chlorimuron-ethyl into soybean field changed the soil microbial community structure significantly.