Merck
  • The protective role of hydrogen sulfide in myocardial ischemia-reperfusion-induced injury in diabetic rats.
The protective role of hydrogen sulfide in myocardial ischemia-reperfusion-induced injury in diabetic rats.
International journal of cardiology (2011-02-15)
Yan Gao, Xiaoyi Yao, Yina Zhang, Weimin Li, Kai Kang, Liang Sun, Xueying Sun
ABSTRACT
Hydrogen sulfide (H(2)S) displays anti-inflammatory and cytoprotective activities to attenuate myocardial ischemia-reperfusion (MIR)-induced injury, but its role in MIR in diabetics is not known. This study was undertaken to investigate whether H(2)S plays a protective role in MIR in diabetic rats. Diabetes was induced by streptozocin in Wistar rats, which were subjected to myocardial ischemia by blocking the left circumflex artery for 30 min, followed by 2h reperfusion. dl-propargylglycine (PAG) and sodium hydrosulfide (NaHS) were administered to the rats to investigate their effects on severity of MIR-induced injury. Diabetic rats had smaller myocardial infarct sizes and higher serum levels of H(2)S (both P < 0.05) than non-diabetics when they underwent MIR. MIR significantly increased the serum level of H(2)S (49.5 ± 7.1 μM), H(2)S-synthesizing activity (7.4 ± 1.6 nmol/mg) and the myocardial infarct size (44.0 ± 7.2%), compared with sham-operated diabetic rats (21.7 ± 2.1 μM, 0.15 ± 0.4 nmol/mg and 1.2 ± 0.4%, respectively). Administration of NaHS increased the H(2)S level (65.8 ± 6.9 μM) and had little effect on H(2)S production activity (6.5 ± 2.2 nmol/mg), while PAG reduced both the H(2)S level (29.2 ± 5.0 μM) and H(2)S-synthesizing activity (2.2 ± 1.8 nmol/mg). NaHS significantly reduced the myocardial infarct size (31.2 ± 4.7%), inhibited the production of lipid peroxidation, MPO activity, and cell apoptosis, and downregulated expression of caspase-3, Fas, FasL, and TNF-α, which had been elevated by MIR, while PAG further increased the myocardial infarct size (58.3 ± 5.9%), and displayed opposite effects. The study indicates that H(2)S may play a protective role in MIR-induced myocardial injury in diabetics by its anti-apoptotic, anti-oxidative and anti-inflammatory activities.
Sigma-Aldrich
DL-Propargylglycine, cystathionine γ-lyase inhibitor
Sigma-Aldrich
L-C-Propargylglycine, ≥99.0% (TLC)