Merck
  • Inter-laboratory calibration of new silver orthophosphate comparison materials for the stable oxygen isotope analysis of phosphates.
Inter-laboratory calibration of new silver orthophosphate comparison materials for the stable oxygen isotope analysis of phosphates.
Rapid communications in mass spectrometry : RCM (2011-02-04)
Stanislaw Halas, Grzegorz Skrzypek, Wolfram Meier-Augenstein, Andrzej Pelc, Helen F Kemp
ABSTRACT
Stable oxygen isotope compositions (δ(18)O values) of two commercial and one synthesized silver orthophosphate reagents have been determined on the VSMOW scale. The analyses were carried out in three different laboratories: lab (1) applying off-line oxygen extraction in the form of CO(2) which was analyzed on a dual inlet and triple collector isotope ratio mass spectrometer, while labs (2) and (3) employed an isotope ratio mass spectrometer coupled to a high-temperature conversion/elemental analyzer (TC/EA) where Ag(3)PO(4) samples were analyzed as CO in continuous flow mode. The δ(18)O values for the proposed new comparison materials were linked to the generally accepted δ(18)O values for Vennemann's TU-1 and TU-2 standards as well as for Ag(3)PO(4) extracted from NBS120c. The weighted average δ(18)O(VSMOW) values for the new comparison materials UMCS-1, UMCS-2 and AGPO-SCRI were determined to be + 32.60 (± 0.12), + 19.40 (± 0.12) and + 14.58 (± 0.13)‰, respectively.
Sigma-Aldrich
Silver phosphate, 98%