- Polychlorinated biphenyls-containing electrical insulating oil contaminated soil treatment with calcium and magnesium peroxides.
Polychlorinated biphenyls-containing electrical insulating oil contaminated soil treatment with calcium and magnesium peroxides.
Chemosphere (2010-12-15)
Anna Goi, Marika Viisimaa, Marina Trapido, Rein Munter
PMID21146854
ABSTRACT
Calcium and magnesium peroxides were applied for the treatment of soil contaminated by polychlorinated biphenyls-containing electrical insulating oil (Aroclor 1016). The removal of PCB-containing electrical insulating oil was achieved with the addition of either calcium peroxide or magnesium peroxide alone and dependent on dosages of the chemical. A 21-d treatment of 60% watered soil with the moderate addition (chemical/oil weight ratio of 0.005/1) of either calcium peroxide or magnesium peroxide resulted in nearly complete (96 ± 2%) oil removal, unsubstantial increase in soil pH and almost no changes in oxygen consumption and dehydrogenase activity, making it suitable for the soil decontamination.