Merck
  • Surface oxygen triggered size change of palladium nano-crystals impedes catalytic efficacy.
Surface oxygen triggered size change of palladium nano-crystals impedes catalytic efficacy.
Chemical communications (Cambridge, England) (2010-12-07)
Jianli Zou, Scott G Stewart, Colin L Raston, K Swaminathan Iyer
ABSTRACT
Palladium nano-crystals increase in size during the initial recycling in Heck cross coupling reactions. We demonstrate that oxygen adsorbed on the surface of palladium nano-crystals plays a pivotal role in driving the ripening. This in turn is associated with a loss in catalytic activity.
Sigma-Aldrich
Butyl acrylate, SAJ first grade, ≥99.0%
Supelco
Butyl acrylate, analytical standard
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Iodobenzene, 98%