Merck
  • Derivatization of 2-chlorophenol with 4-amino-anti-pyrine: a novel method for improving the selectivity of molecularly imprinted solid phase extraction of 2-chlorophenol from water.
Derivatization of 2-chlorophenol with 4-amino-anti-pyrine: a novel method for improving the selectivity of molecularly imprinted solid phase extraction of 2-chlorophenol from water.
Talanta (2010-11-30)
Amjad H El-Sheikh, Rasha W Al-Quse, Musa I El-Barghouthi, Fida'a S Al-Masri
ABSTRACT
Molecularly imprinted polymer (MIP) may not selectively recognize small template of limited number of functional groups, such as 2-chlorophenol (2-CP). In this work, a novel method was proposed to improve the recognition ability of the molecularly imprinted solid phase extraction (MISPE) of 2-CP from environmental waters. This was achieved by derivatization of 2-CP with 4-amino-anti-pyrine (4-AAP) to enlarge its molecular size and add more binding sites. For that purpose, two MISPE methods of 2-CP were developed. In method 1, a polymer imprinted with 2-CP was used as the extracting sorbent but it suffered from low selectivity and high detection limit of 2-CP (7.10 ng L(-1)). In method 2, a polymer imprinted with 4-AAP derivatized 2-CP (2-CP-4-AAP) was used as the extracting sorbent. Prior to loading the water sample it was subjected to a simple derivatization procedure with 4-AAP. Method 2 showed high recognition ability/selectivity towards 2-CP-4-AAP with lower detection limit of 0.05 ng L(-1) for 2-CP-4-AAP. Method 2 was able to detect the presence of 2-CP-4-AAP in unspiked real water samples and almost full spike recovery was achieved.
Sigma-Aldrich
4-Aminoantipyrine, puriss. p.a., reag. Ph. Eur., ≥99%
Sigma-Aldrich
4-Aminoantipyrine, reagent grade
Sigma-Aldrich
4-Aminoantipyrine, ≥99.0%, JIS special grade
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%