Merck
  • Amidation of alcohols with nitriles under solvent-free conditions using molecular iodine as a catalyst.
Amidation of alcohols with nitriles under solvent-free conditions using molecular iodine as a catalyst.
Journal of oleo science (2010-10-26)
Yoshio Kasashima, Atsushi Uzawa, Kahoko Hashimoto, Yu Yokoyama, Takashi Mino, Masami Sakamoto, Tsutomu Fujita
ABSTRACT
The reactions of alcohols with nitriles under solvent-free conditions, using molecular iodine as a catalyst, were investigated. The reaction of 1-phenylethanol with propanenitrile produced the amide N-(1-phenylethyl)propanamide, by dehydration and tautomerization, in 71% yield, under the following conditions: temperature=90°C, alcohol:iodine molar ratio=1:0.2, alcohol:nitrile molar ratio=1:5, and reaction time=5 h. The amidation reactivity depended on the stability of the cationic intermediate formed from the alcohol. The reaction of (-)-borneol with benzonitrile produced a racemic amide in 83% yield.
Sigma-Aldrich
α-Methylbenzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
1-Phenylethanol, 98%