Merck
  • Azide-specific labeling of biomolecules by Staudinger-Bertozzi ligation phosphine derivatives of fluorescent probes suitable for single-molecule fluorescence spectroscopy.
Azide-specific labeling of biomolecules by Staudinger-Bertozzi ligation phosphine derivatives of fluorescent probes suitable for single-molecule fluorescence spectroscopy.
Methods in enzymology (2010-06-29)
Anirban Chakraborty, Dongye Wang, Yon W Ebright, Richard H Ebright
ABSTRACT
We describe the synthesis of phosphine derivatives of three fluorescent probes that have a brightness and photostability suitable for single-molecule fluorescence spectroscopy and microscopy: Alexa488, Cy3B, and Alexa647. In addition, we describe procedures for use of these reagents in azide-specific, bioorthogonal labeling through Staudinger-Bertozzi ligation, as well as procedures for the quantitation of labeling specificity and labeling efficiency. The reagents and procedures of this report enable chemoselective, site-selective labeling of azide-containing biomolecules for single-molecule fluorescence spectroscopy and microscopy.
Sigma-Aldrich
8-(6-Aminohexyl)aminoadenosine 3′:5′-cyclic monophosphate, ~98%