Merck
Upper gastrointestinal hemorrhage caused by superwarfarin poisoning.
World journal of gastroenterology (2010-04-01)
Shu Lei Zhao, Peng Li, Ming Ji, Ye Zong, Shu Tian Zhang
ABSTRACT
Superwarfarins are a class of rodenticides. Gastrointestinal hemorrhage is a fatal complication of superwarfarin poisoning, requiring immediate treatment. Here, we report a 55-year-old woman with tardive upper gastrointestinal hemorrhage caused by superwarfarin poisoning after endoscopic cold mucosal biopsy.